Anti-CD2 Rat Monoclonal Antibody

GTX74805
89365-586EA 244.71 USD
89365-586
Anti-CD2 Rat Monoclonal Antibody
Antibodies
Rat monoclonal antibody [AT37] to CD2

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR