Anti-ITGB2 Mouse Monoclonal Antibody

Supplier: Genetex

GTX78280
89366-580EA 381.15 USD InStock
89366-580
Anti-ITGB2 Mouse Monoclonal Antibody
Antibodies
Monoclonal Antibody to CD18 (Human) Azide Free

Flow Cytometry; Immunoprecipitation; Western Blotting: Recommended dilution:2 _g/ml, Positive control:Kg-1a human leukemia cell lysate, Negative control:JURKAT human leukemia T-cell lysate, Sample preparation:buffer with laurylmaltoside, 2 x non-reducing SDS, Application note:Non-reducing conditions. SDS-PAGE (6% separating gel, 4% stacking gel). Recommended secondary antibody - anti mouse IgG/HRP, dilution 1:2000, 60 min on vertical incubator. Immunohistochemistry (frozen section); Functional Application: The antibody induces high-affinity conformation of LFA-1 complex.

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR