Anti-CD175 Mouse Monoclonal Antibody [clone: BRIC111]

Supplier: GeneTex

GTX75684
89365-872EA 517.32 USD
89365-872
Anti-CD175 Mouse Monoclonal Antibody [clone: BRIC111]
Antibodies
Tn is a cryptantigen which was recently designated CD175 at the 7th Leucocyte Typing Workshop. Tn antigen is not expressed on normal haemopoietic cells but exposure of the Tn is associated with polyagglutination.

Type: Primary
Antigen: CD175
Clonality: Monoclonal
Clone: BRIC111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR