Anti-CD172A Mouse Monoclonal Antibody [clone: MRC OX-41]

Supplier: GeneTex
GTX76052
89365-968EA 625.2 USD
89365-968
Anti-CD172A Mouse Monoclonal Antibody [clone: MRC OX-41]
Antibodies
Mouse Monoclonal antibody [MRC OX-41] to SIRP alpha

For FACS: Use at an assay dependent dilution. For IF: Use at an assay dependetn dilution. For ICC: Use at an assay dependent dilution. For RIA (indirect): Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD172A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR