Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSGL-1 Mouse Monoclonal Antibody [clone: 3E2.25.5(PL1)]

Supplier: Genetex
GTX19793
89360-694
Anti-PSGL-1 Mouse Monoclonal Antibody [clone: 3E2.25.5(PL1)]
Selectin P ligand, or P-selectin glycoprotein ligand (PSGL-1), is the high affinity counter-receptor for P-selectin on myeloid cells and stimulated T lymphocytes. As such, it plays a critical role in the tethering of these cells to activated platelets or endothelia expressing P-selectin.

FACS: Use at an assay dependent dilution. IHC-Fr: 1/200. IHC-P: 1/200. WB: Use at an assay dependent dilution. Predicted molecular weight: 43 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSGL-1
Clonality: Monoclonal
Clone: 3E2.25.5(PL1)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty