Anti-CD16 Mouse Monoclonal Antibody [clone: MEM-154]

Supplier: Genetex

GTX78279
89366-578EA 367 USD
89366-578
Anti-CD16 Mouse Monoclonal Antibody [clone: MEM-154]
Antibodies
Monoclonal Antibody to CD16 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry. Suggested working dilution is 1:500. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR