CD16 is a 50-65kD cell surface molecule which exists in two forms - a transmembranous form expressed by NK cells and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells.
FACS: Use at an assay dependent concentration. IP: Use at an assay dependent concentration. WB: Use at an assay dependent concentration. Use under heat-denatured non-reducing conditions (no reducing agent). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: MEM-154
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human