Anti-CD16 Mouse Monoclonal Antibody [clone: LNK16]

Supplier: Genetex

GTX18234
89351-450EA 403.98 USD InStock
89351-450
Anti-CD16 Mouse Monoclonal Antibody [clone: LNK16]
Antibodies
CD16 is a cell surface molecule which exists in two forms - a transmembranous form expressed by NK cells and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells.

FACS: Use 20?l for 100?l human whole blood cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR