Anti-CD14 Mouse Monoclonal Antibody [clone: B365.1 (B-A8)]

Supplier: GeneTex
GTX72062
89352-944EA 322.1 USD
89352-944
Anti-CD14 Mouse Monoclonal Antibody [clone: B365.1 (B-A8)]
Antibodies
Mouse monoclonal [B365.1 (B-A8)] to CD14

Immunofluorescence 1:50-1:100 .

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: B365.1 (B-A8)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR