Anti-ENG Mouse Monoclonal Antibody [clone: SN6]

Supplier: GeneTex

GTX75142
89353-340EA 517.19 USD
89353-340
Anti-ENG Mouse Monoclonal Antibody [clone: SN6]
Antibodies
Mouse monoclonal antibody [SN6] to CD105

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR