Anti-CCRL2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108151
89355-316EA 427.1 USD
89355-316
Anti-CCRL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CCRL2 (chemokine (C-C motif) receptor-like 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CCRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR