Anti-CCR11 Goat Polyclonal Antibody

Supplier: GeneTex
GTX21660
89361-052EA 611.88 USD
89361-052
Anti-CCR11 Goat Polyclonal Antibody
Antibodies
Receptors for the C-C chemokine family include CCR-1, CCR-2A, CCR-3, CCR-4, CCR-5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. CCR11 in particular is expressed in the heart, lung, and small intestine. It binds members of the monocyte chemoattractant family including MCP-1, MCP-2, and MCP-4.

ELISA: Use at a dilution of 1/75000. ICC: Use at a dilution of 1/500. IHC-P: Use at a dilution of 1/250. WB: Use at a dilution of 1/1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CCR11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR