Anti-CBP Mouse Monoclonal Antibody [clone: F4]

Supplier: GeneTex
GTX30750
89352-736EA 625.2 USD
89352-736
Anti-CBP Mouse Monoclonal Antibody [clone: F4]
Antibodies
Mouse monoclonal [F4] to CBP

For IF: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user

Type: Primary
Antigen: CBP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR