Anti-CBL Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101352
89349-000EA 582.76 USD
89349-000
Anti-CBL Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CBL (Cas-Br-M (murine) ecotropic retroviral transforming sequence)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:50-1:200. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR