Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

GTX77004
89353-426EA 152.47 USD
89353-426
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
FITC-conjugated Goat anti-Cat IgG Polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR