Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP3 Mouse Monoclonal Antibody [clone: CPP32 4-1-18]

Supplier: Genetex

GTX30246
89362-902EA 431.12 USD InStock
89362-902
Anti-CASP3 Mouse Monoclonal Antibody [clone: CPP32 4-1-18]
Antibodies
Mouse monoclonal ( CPP32 4 - 1 - 18 ) to human Caspase 3

For WB: Use at a dilution of 1:500 - 1:1,000. May be used for Western analysis where a band is seen at ~32 kDa for the inactive form of Caspase 3 and ~17-22 kDa for the active form of Caspase 3. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: CPP32 4-1-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty