Anti-TNNT2 Mouse Monoclonal Antibody [clone: 2F3]

Supplier: GeneTex
GTX10216
89350-708EA 730.23 USD
89350-708
Anti-TNNT2 Mouse Monoclonal Antibody [clone: 2F3]
Antibodies
Mouse monoclonal [2F3] to cardiac Troponin T

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 35.4 kDa. Can be paired for ELISA with GTX10223 Mouse monoclonal [7G7] to cardiac Troponin T . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Dog, Pig, Rat, Cat, Fish, Goat, Rabbit, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR