Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-TNNI3 Mouse Monoclonal Antibody [clone: 16A12]

Supplier: Genetex

GTX10227
89350-722EA 246.42 USD InStock
89350-722
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 16A12]
Antibodies
Mouse monoclonal [16A12] to cardiac Troponin I

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 16A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty