Anti-TNNC1 Mouse Monoclonal Antibody [clone: 7B9]

Supplier: GeneTex
GTX28286
89352-404EA 630.94 USD
89352-404
Anti-TNNC1 Mouse Monoclonal Antibody [clone: 7B9]
Antibodies
Mouse monoclonal [7B9] to cardiac Troponin C

AP: Use at an assay dependent concentration. ELISA: Use at an assay dependent concentration. IA: Use at an assay dependent concentration. WB: Use at an assay dependent concentration. Can be paired for ELISA with GTX28285 Mouse monoclonal [1A2] to cardiac Troponin C . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNNC1
Clonality: Monoclonal
Clone: 7B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR