Anti-TNNC1 Mouse Monoclonal Antibody [clone: 12G3]

GTX10245
89350-740EA 341.54 USD
89350-740
Anti-TNNC1 Mouse Monoclonal Antibody [clone: 12G3]
Mouse monoclonal [12G3] to cardiac Troponin C

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 19 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNNC1
Clonality: Monoclonal
Clone: 12G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR