Anti-CAST Mouse Monoclonal Antibody [clone: CSL1 5]

Supplier: GeneTex
GTX12217
89359-278EA 581.61 USD
89359-278
Anti-CAST Mouse Monoclonal Antibody [clone: CSL1 5]
Antibodies
Calpastatin is an endogenous calpain (calcium-dependent cysteine protease) inhibitor. It consists of an N-terminal domain L and four repetitive calpain-inhibition domains (domains 1-4), and it is involved in the proteolysis of amyloid precursor protein. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation. The protein is also thought to affect the expression levels of genes encoding structural or regulatory proteins.

ELISA: Use at an assay dependent dilution. Pair with GTX10320 for muscle calpastatin. WB: Use at an assay dependent dilution. Predicted molecular weight: 76.5 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CAST
Clonality: Monoclonal
Clone: CSL1 5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR