Anti-CEBPE Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109155
89355-794EA 1199.11 USD
89355-794
Anti-CEBPE Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to C/EBP epsilon (CCAAT/enhancer binding protein (C/EBP), epsilon)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: CEBPE
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR