Mouse monoclonal antibody [1H8] to C-Peptide
Competitive ELISA or RIA detection of C-peptide in human serum without or after immunospecific removal of proinsulin (with solid-phased GTX28299 (1D6) to human proinsulin)Construction of a sensitive two-site sandwich EIA/ RIA immunosorbent assays for the measurement of total (including proinsulin convertion intermediates) human proinsulin in serum. GTX28298 (2B7) used in conjunction with this antibody would be suitable. Specific immunosorbtional (with solid-phase 1H8) removal of total proinsulin and C-peptide molecules from serum samples before the measurement of human insulin
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human