Anti-BRUCellA ABORTUS Mouse Monoclonal Antibody

GTX22022
89351-736EA 299.5 USD
89351-736
Anti-BRUCellA ABORTUS Mouse Monoclonal Antibody
Brucella abortus is the causative agent of brucellosis.

Type: Primary
Antigen: BRUCellA ABORTUS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR