Anti-GDF2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108417
89355-512EA 557.47 USD
89355-512
Anti-GDF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BMP9 (growth differentiation factor 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GDF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR