Anti-BMP5 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10858
89355-596EA 606.93 USD
89355-596
Anti-BMP5 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to BMP5

For ELISA: Use at a concentration of 0.5-1.0 ?g/ml. For ICC: Use at a concentration of 1-3 ?g/ml. For IHC: Use at a concentration of 5-10 ?g/ml. For WB: Use at a concentration of 0.1-0.2 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: BMP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR