Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 6F9]

Supplier: GeneTex
GTX26300
89361-870EA 519.91 USD
89361-870
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 6F9]
Antibodies
Mouse monoclonal antibody [6F9] to beta Catenin

  • ICC: Use at an assay dependant dilution. IF: Use at a dilution of 1/1000. WB: Use at a dilution of 1/1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR