Anti-B2M Mouse Monoclonal Antibody [clone: B2M-02]

Supplier: Genetex

GTX78408
89366-632EA 353.77 USD InStock
89366-632
Anti-B2M Mouse Monoclonal Antibody [clone: B2M-02]
Antibodies
Monoclonal Antibody to beta2-Microglobulin Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: B2M-02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR