Anti-ABCG2 Mouse Monoclonal Antibody [clone: BXP-34]

Supplier: GeneTex
GTX23379
89352-004EA 611.89 USD
89352-004
Anti-ABCG2 Mouse Monoclonal Antibody [clone: BXP-34]
Antibodies
Mouse Monoclonal antibody [BXP-34] to ABCG2

This antibody does not work in Western blot. Optimal dilutions / concentrations should be determined by the end user.

Type: Primary
Antigen: ABCG2
Clonality: Monoclonal
Clone: BXP-34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR