Anti-AU5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23405
89352-016EA 591 USD
89352-016
Anti-AU5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AU5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: TDFYLK
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR