Anti-ATG4D Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX106114
89354-506EA 426.96 USD
89354-506
Anti-ATG4D Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATG4D (ATG4 autophagy related 4 homolog D (S. cerevisiae))

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ATG4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR