Anti-ATF1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100062
89348-180EA 557.66 USD
89348-180
Anti-ATF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATF1 (activating transcription factor 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR