Anti-ARPC1A Goat Polyclonal Antibody

Supplier: Genetex

GTX22752
89361-320EA 430.61 USD
89361-320
Anti-ARPC1A Goat Polyclonal Antibody
Antibodies
Goat polyclonal to ARPC1A

ELISA: Peptide ELISA (antibody detection limit dilution 1:32,000). Western Blot: No signal obtained yet but low background observed in Human Testis and A431 lysates at up to 2ug/ml.

Type: Primary
Antigen: ARPC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR