Anti-AP3D1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX23705
89367-564EA 799.9 USD
89367-564
Anti-AP3D1 Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Ap3D1

Recommended Starting Dilutions:WB: Use at 1:500 - 1:2,000ELISA: Use at 1:5,000 - 1:20,000IP: Use at 1:100

Type: Primary
Antigen: AP3D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR