Anti-PRKAA2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23760
89352-054EA 611 USD
89352-054
Anti-PRKAA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AMPK alpha 2

For IP: Use at a concentration of 2 - 8 ?g/ml. For WB: Use at a dilution of 0.2 - 2?g/ml. Predicted molecular weight: 63 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR