Anti-IGFALS Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102294
89349-538EA 582.76 USD
89349-538
Anti-IGFALS Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ALS (insulin-like growth factor binding protein, acid labile subunit)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IGFALS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR