Anti-TUBA1 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX18247
89360-440EA 611.88 USD
89360-440
Anti-TUBA1 Mouse Monoclonal Antibody
Antibodies
Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement and in the cytoskeleton. Tubulin is a heterodimer which consists of alpha tubulin and beta tubulin.

Type: Primary
Antigen: TUBA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR