Anti-ALDOA Goat Polyclonal Antibody

Supplier: GeneTex
GTX26190
89361-852EA 764.87 USD
89361-852
Anti-ALDOA Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALDOA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR