Anti-AKT3 Rabbit Polyclonal Antibody

GTX113311
89357-462EA 376.43 USD
89357-462
Anti-AKT3 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to AKT3 (v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR