Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Monoclonal Antibody [clone: PKB-175]

Supplier: Genetex

GTX10438
89350-766EA 325.12 USD InStock
89350-766
Anti-AKT1 Mouse Monoclonal Antibody [clone: PKB-175]
Antibodies
Mouse monoclonal antibody [PKB-175] to Akt1

For WB: Use at a dilution of 1:5,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: PKB-175
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty