Anti-ADCK4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111571
89356-780EA 582.56 USD
89356-780
Anti-ADCK4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ADCK4 (aarF domain containing kinase 4)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ADCK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR