Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDAP1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX22108
89361-160EA 217.48 USD InStock
89361-160
Anti-PDAP1 Rabbit Polyclonal Antibody
Antibodies
Acid phosphatase 1 soluble antibody Acid phosphatase of erythrocyte antibody Adipocyte acid phosphatase antibody Cytoplasmic phosphotyrosyl protein phosphatase antibody HAAP antibody Low molecular weight phosphotyrosine protein phosphatase antibody PAP1 antibody PAP2 antibody Protein tyrosine phosphatase antibody PTPase antibody Purple acid phosphatase antibody Red cell acid phosphatase 1 antibody

Suggested working dilution(s):Standard sandwich ELISA against 1.0µg Potato Acid Phosphatase - 1:20,000 to 1:80,000Optimal titers for other applications should be determined by the researcher.

Type: Primary
Antigen: PDAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty