Anti-ACE Mouse Monoclonal Antibody [clone: M4110421]

Supplier: GeneTex
GTX22092
89361-150EA 588.41 USD
89361-150
Anti-ACE Mouse Monoclonal Antibody [clone: M4110421]
Antibodies
Angiotensin Converting enzyme is involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system.

ELISA: Use at an assay dependent dilution. FACS: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR