Anti-ACE Mouse Monoclonal Antibody [clone: i2H5]

GTX11736
89358-764EA 379.2 USD
89358-764
Anti-ACE Mouse Monoclonal Antibody [clone: i2H5]
ACE converts angiotensin I to angiotensin II by release of the terminal His-Leu, this results in an increase of the vasoconstrictor activity of angiotensin. Also able to inactivate bradykinin, a potent vasodilatator. ACE exists in two forms, a 170KD somatic form and a 90KD germinal form. The somatic form is expressed by endothelial cells (especially those of lung capillaries and arterioles), epithelial cells (especially in proximal renal tubules and small intestine), by some neuronal cells and variably by some macrophages and T lymphocytes. The germinal form is expressed by spermatozoa.

Recommended Starting Dilutions:Flow Cytometry: Neat - 1/10Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR