This protein is a medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. This enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Clinical phenotypes are associated with ACADM hereditary deficiency.
Approx 45kDa band observed in Human Kidney and Heart lysates (calculated MW of 46.6kDa according to NP_000007.1). Recommended for use at 0.01-0.1µg/ml. In paraffin embedded Human Liver shows mitochondrial staining in hepatocytes. Recommended concentration, 3-5µg/ml. This antibody is expected to recognise both reported isoforms.
Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human