Anti-5HT1B Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX13896
89359-888EA 462.26 USD
89359-888
Anti-5HT1B Rabbit Polyclonal Antibody
Antibodies
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.

For WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 40-41 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: 5HT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR