Anti-FTCD Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25820
89361-752EA 602.26 USD
89361-752
Anti-FTCD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to 58K Golgi protein - Golgi Marker

WB: Use at a dilution of 1/500 - 1/1000. Detects a band of approximately 58 kDa (predicted molecular weight: 58 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user. We are currently arranging to have this antibody tested in immunofluoresence.

Type: Primary
Antigen: FTCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR