Anti-TNFA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX74120
89365-388EA 811.37 USD
89365-388
Anti-TNFA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TNF alpha

Recommended Starting Dilutions: ELISA: 0.5 - 2.0 ug/ml Functional Assays: 0.07 - 0.1 ug/ml Western Blotting: 0.1 - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR