Anti-TGFA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27293
89362-532EA 650.28 USD
89362-532
Anti-TGFA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to TGF alpha

  • ELISA: Use at a concentration of 0.5 μg/ml. Allows detection of 0.2 - 0.4 ng/well of recombinant hTGF-a. Neut: Use at a concentration of 0.1 - 0.15 μg/ml to yield one-half maximal inhibition [ND50] of the biological activity of hTGF-a (2 ng/ml). WB: Use at a concentration of 0.1 - 0.2 μg/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR