Anti-SLUG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30813
89363-004EA 602.93 USD
89363-004
Anti-SLUG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SLUG

Recommended Starting Dilutions:Western Blotting: 0.5 - 2.0 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: SLUG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR